Assay Details
| Gene Symbol: LOC101927432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040919.1 | - | 1 - 2 | 152 | 71 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM920490.1 | - | 168 | Chr. - - [-] on Build | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TMEM108 antisense RNA 1 | TMEM108-AS1 | -127.2kb | Sense | Chr. 3 - 133246448 - 133257121 on NCBI Build GRCh38 | |
| Downstream | BFSP2 antisense RNA 1 | BFSP2-AS1 | 16.4kb | Sense | Chr. 3 - 133426402 - 133491146 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927432
Genomic Map