Assay Details
| Gene Symbol: LOC101927432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040919.1 | - | 4 - 5 | 490 | 85 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM920490.1 | - | 506 | Chr. - - [-] on Build | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TMEM108 antisense RNA 1 | TMEM108-AS1 | -109.3kb | Sense | Chr. 3 - 133246448 - 133257121 on NCBI Build GRCh38 | |
| Downstream | beaded filament structural protein 2 | BFSP2 |
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN |
31.4kb | Anti-sense | Chr. 3 - 133397882 - 133475212 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927432
Genomic Map