Assay Details
| Gene Symbol: LOC100129603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038729.1 | - | 3 - 4 | 274 | 150 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483511 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269653 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483663 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269605 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269596 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483664 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269660 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269664 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483658 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269595 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269768 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15902644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269762 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25483665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05269748 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3711619 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56841 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P17828110 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P01645688 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37906980 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17828110 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01645688 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173655 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 85497 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 32412 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 68760 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645688 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17828061 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P25343642 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645717 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645711 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645702 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37906980 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17828056 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15962306 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645714 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17828110 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37906982 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01645709 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P25343985 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563324_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000457522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000457522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000458648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | ENST00000458648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000402115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000403226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000404626.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000457522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000457522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000458648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | ENST00000458648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010092.hg.1 | NR_126024.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 10694991 | - | - |

LOC100129603
Genomic Map