Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105580.2 | NP_001099050.1 | 5 - 6 | 642 | 81 | |
| NR_047685.1 | 5 - 6 | 642 | 81 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001600.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028788.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028788.hg.1 | ENST00000472788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028788.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028786.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028786.hg.1 | ENST00000472788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028786.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028787.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028787.hg.1 | ENST00000472788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028787.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187635.hg.1 | TC0300011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187634.hg.1 | TC0300011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187636.hg.1 | TC0300011802.hg.1 | Clariom D TC |

GABRR3
Genomic Map