Assay Details
Gene Symbol: LIMD1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033947.1 | 1 - 2 | 503 | 111 | ||
GenBank mRNA | AK026206.1 | - | 1 - 2 | 504 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001349.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 553177 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 526352 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 79294 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00696200 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00696200 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00696201 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16209989 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36249830 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P408005 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00696200 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8161944 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8086539 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117080 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8097475 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117079 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8079334 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8161943 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8097474 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8086538 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 233541_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR03023006.hg.1 | TC03001349.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023006.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023006.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023002.hg.1 | TC03001349.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023002.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023004.hg.1 | TC03001349.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023004.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023004.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023005.hg.1 | TC03001349.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023005.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023005.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179584.hg.1 | TC0300010928.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179580.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179580.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179580.hg.1 | TC0300010928.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179585.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179585.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179585.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179585.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179585.hg.1 | TC0300010928.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179583.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179583.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179583.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179583.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179583.hg.1 | TC0300010928.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000427644.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | ENST00000429798.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010928.hg.1 | NR_033947.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 233541_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 233541_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 233541_at7 | - | - |
Human Genome U95 Set | 53220_at | - | - |
Human Genome U95 Set | 53220_PM_at | - | - |
Human Genome U133 Set | 233541_PM_at | - | - |
Human Genome U133 Set | 233541_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020968 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020968 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 102831 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 188439 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1684266 | AK026206.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1684266 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684266 | AK026206.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1684266 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658748 | AK026206.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658748 | BC047037.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658748 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658748 | AJ132408.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1658748 | AK021508.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752415 | AJ132408.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752415 | BC027712.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752415 | NM_014240.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1684266 | AK026206.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1684266 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658748 | AK026206.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658748 | BC047037.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658748 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658748 | AJ132408.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1658748 | AK021508.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684266 | AK026206.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684266 | NR_033947.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752415 | AJ132408.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752415 | BC027712.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752415 | NM_014240.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1684266 | AK026206.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1684266 | NR_033947.1 | human_transcriptome |
