Assay Details
| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004326.4 | NP_001004326.4 | 2 - 3 | 517 | 71 | |
| NR_033396.1 | 2 - 3 | 517 | 71 | |||
| XM_011545043.2 | XP_011543345.1 | 2 - 3 | 517 | 71 | ||
| XM_017017774.1 | XP_016873263.1 | 2 - 3 | 517 | 71 | ||
| GenBank mRNA | AK131327.1 | - | 2 - 3 | 519 | 71 | |
| BC146850.1 | - | 2 - 3 | 509 | 9056778 | 71 | |
| DY654625.1 | - | 2 - 3 | 159 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000621.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11007710.hg.1 | ENST00000525437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007710.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007710.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007710.hg.1 | TC11000621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007711.hg.1 | ENST00000525437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007711.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007711.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007711.hg.1 | TC11000621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007709.hg.1 | ENST00000525437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007709.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007709.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007709.hg.1 | TC11000621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007712.hg.1 | ENST00000525437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007712.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007712.hg.1 | TC11000621.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157717.hg.1 | TC1100008006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157718.hg.1 | TC1100008006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | ENST00000534471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008006.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157719.hg.1 | TC1100008006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000529062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | ENST00000530038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157720.hg.1 | TC1100008006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | ENST00000525264.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | ENST00000525437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | NM_001004326.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | NR_033396.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100157715.hg.1 | TC1100008006.hg.1 | Clariom D TC |

SLC22A20
Genomic Map