Assay Details
| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010874.4 | NP_001010874.2 | 1 - 2 | 342 | 154 | |
| XM_005265663.2 | XP_005265720.1 | 1 - 2 | 342 | 154 | ||
| XM_005265664.2 | XP_005265721.1 | 1 - 2 | 342 | 154 | ||
| XM_005265665.3 | XP_005265722.1 | 1 - 2 | 342 | 154 | ||
| XM_017007959.1 | XP_016863448.1 | 1 - 2 | 342 | 154 | ||
| XM_017007960.1 | XP_016863449.1 | 1 - 2 | 342 | 154 | ||
| GenBank mRNA | AK290606.1 | - | 1 - 2 | 338 | 154 | |
| AL833108.1 | - | 1 - 2 | 343 | 154 | ||
| BC072463.1 | - | 1 - 2 | 342 | 4245690 | 154 | |
| BX647590.1 | - | 1 - 2 | 279 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001242.hg.1
Set Membership: | 
		   
					
						
							 | 
				
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Affymetrix
					
					
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04016871.hg.1 | TC04001242.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016871.hg.1 | ENST00000381210.3 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016871.hg.1 | ENST00000507440.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016871.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016870.hg.1 | TC04001242.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016870.hg.1 | ENST00000381210.3 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016870.hg.1 | ENST00000507440.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016870.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016869.hg.1 | TC04001242.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016869.hg.1 | ENST00000381210.3 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016869.hg.1 | ENST00000507440.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR04016869.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173745.hg.1 | TC0400010853.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173743.hg.1 | TC0400010853.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173746.hg.1 | TC0400010853.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000381210.7 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000507440.5 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000509536.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | ENST00000511356.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | NM_001010874.4 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0400173744.hg.1 | TC0400010853.hg.1 | Clariom D TC | 

TECRL
Genomic Map