Assay Details
Gene Symbol: MRGPRF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120541.1 | 1 - 2 | 193 | 69 | ||
GenBank mRNA | BC039516.1 | - | 1 - 2 | 116 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160632.hg.1 | TC1100008217.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160629.hg.1 | TC1100008217.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008217.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000538407.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000569428.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | ENST00000569432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | NR_120541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100160630.hg.1 | TC1100008217.hg.1 | Clariom D TC |
