Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004281.hg.1 |
TC04000302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004281.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004281.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007518.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151356.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151356.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151356.hg.1 |
ENST00000514957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151356.hg.1 |
NR_040106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151356.hg.1 |
TC0400007518.hg.1 |
Clariom D TC |