| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004066.hg.1 |
TC07000267.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004066.hg.1 |
ENST00000265523.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004066.hg.1 |
ENST00000402924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004066.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004066.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000424330.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000453612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007352.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000265523.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000402924.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
ENST00000486984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
NM_000712.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
NM_001253823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150946.hg.1 |
TC0700007352.hg.1 |
Clariom D TC |