Assay Details
| Gene Symbol: MADCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC136408.1 | - | 1 - 2 | 98 | 9020396 | 140 |
| BC137506.1 | - | 1 - 2 | 98 | 9021506 | 140 | |
| BC137507.1 | - | 1 - 2 | 98 | 9021507 | 140 | |
| BC144065.1 | - | 1 - 2 | 98 | 9052585 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC136408.1, BC137506.1, BC137507.1, BC144065.1 transcripts.
Set Membership: |
|

MADCAM1
Genomic Map