| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061429.hg.1 |
TC01004071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061429.hg.1 |
ENST00000294753.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061429.hg.1 |
ENST00000366498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061429.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061427.hg.1 |
TC01004071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061427.hg.1 |
ENST00000294753.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061427.hg.1 |
ENST00000366498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061427.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061428.hg.1 |
TC01004071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061428.hg.1 |
ENST00000294753.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061428.hg.1 |
ENST00000366498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061428.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000461277.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000462139.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018097.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236211.hg.1 |
TC0100018097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100236213.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236213.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236213.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236213.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236213.hg.1 |
TC0100018097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000294753.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000477903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
ENST00000478225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
NM_032752.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236212.hg.1 |
TC0100018097.hg.1 |
Clariom D TC |