Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032752.1 | NP_116141.1 | 5 - 6 | 1109 | 106 | |
| XM_005273330.3 | XP_005273387.1 | 6 - 7 | 2600 | 106 | ||
| XM_011544312.1 | XP_011542614.1 | 6 - 7 | 2600 | 106 | ||
| XM_017002589.1 | XP_016858078.1 | 6 - 7 | 2600 | 106 | ||
| XM_017002590.1 | XP_016858079.1 | 6 - 7 | 2600 | 106 | ||
| XM_017002591.1 | XP_016858080.1 | 6 - 7 | 2553 | 106 | ||
| GenBank mRNA | AK098030.1 | - | 1 - 2 | 222 | 106 | |
| AK310669.1 | - | 1 - 2 | 231 | 106 | ||
| BC007263.1 | - | 5 - 6 | 1109 | 3051320 | 106 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01004071.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
protein self-association
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
protein self-association
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01061422.hg.1 | TC01004071.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061422.hg.1 | ENST00000294753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061422.hg.1 | ENST00000366498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061422.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061423.hg.1 | TC01004071.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061423.hg.1 | ENST00000294753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061423.hg.1 | ENST00000366498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061423.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061415.hg.1 | TC01004071.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061415.hg.1 | ENST00000294753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061415.hg.1 | ENST00000366498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061415.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061415.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061419.hg.1 | TC01004071.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061419.hg.1 | ENST00000294753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061419.hg.1 | ENST00000366498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061419.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061424.hg.1 | TC01004071.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061424.hg.1 | ENST00000294753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061424.hg.1 | ENST00000366498.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01061424.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236200.hg.1 | TC0100018097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000462139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018097.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236205.hg.1 | TC0100018097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236203.hg.1 | TC0100018097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236206.hg.1 | TC0100018097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000294753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000461277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000477903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | ENST00000478225.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | NM_032752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100236207.hg.1 | TC0100018097.hg.1 | Clariom D TC |

ZNF496
Genomic Map