Assay Details
| Gene Symbol: CBARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152769.2 | NP_689982.3 | 1 - 2 | 241 | 156 | |
| XM_017026556.1 | XP_016882045.1 | 1 - 2 | 215 | 156 | ||
| GenBank mRNA | AW163263.1 | - | 1 - 2 | 242 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000998.hg
Additional Information:
This assay is in the 5'UTR region of the NM_152769.2,XM_017026556.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19013648.hg.1 | ENST00000215376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013648.hg.1 | ENST00000382477.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013648.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013648.hg.1 | TC19000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013651.hg.1 | ENST00000215376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013651.hg.1 | ENST00000382477.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013651.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013651.hg.1 | TC19000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013649.hg.1 | ENST00000215376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013649.hg.1 | ENST00000382477.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013649.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013649.hg.1 | TC19000998.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013650.hg.1 | ENST00000215376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013650.hg.1 | ENST00000382477.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013650.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013650.hg.1 | TC19000998.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | TC1900009137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172669.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | TC1900009137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172671.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | TC1900009137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172674.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | TC1900009137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172667.hg.1 | NM_152769.2 | Refseq |

CBARP
Genomic Map