Assay Details
| Gene Symbol: CBARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026555.1 | XP_016882044.1 | 9 - 10 | 1530 | 125 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |

CBARP
Genomic Map