Assay Details
| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016186.2 | NP_057270.1 | 1 - 2 | 420 | 93 | |
| XM_017021353.1 | XP_016876842.1 | 2 - 3 | 371 | 93 | ||
| GenBank mRNA | AF181467.1 | - | 1 - 2 | 420 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001464.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF181467.1,NM_016186.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14016484.hg.1 | ENST00000393096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016484.hg.1 | ENST00000554723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016484.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016484.hg.1 | TC14001464.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174343.hg.1 | TC1400010042.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010042.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000261994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000554173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174342.hg.1 | TC1400010042.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | ENST00000393096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | ENST00000554723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | NM_016186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400174345.hg.1 | TC1400010042.hg.1 | Clariom D TC |

SERPINA10
Genomic Map