Assay Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138418.2 | NP_612427.2 | 4 - 5 | 560 | 83 | |
| GenBank mRNA | AK054811.1 | - | 2 - 3 | 1956 | 83 | |
| BC001912.1 | - | 4 - 5 | 560 | 3546959 | 83 | |
| BC007346.2 | - | 3 - 4 | 352 | 83 | ||
| BC111391.1 | - | 4 - 5 | 462 | 6457301 | 83 | |
More Information
Set Membership: |
|

MCRIP2
Genomic Map