Assay Details
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198066.3 | NP_932332.1 | 3 - 4 | 466 | 121 | |
XM_005268012.2 | XP_005268069.1 | 4 - 5 | 500 | 121 | ||
XM_006720238.3 | XP_006720301.1 | 3 - 4 | 599 | 121 | ||
GenBank mRNA | AK090577.1 | - | 3 - 4 | 399 | 121 | |
BC012179.1 | - | 3 - 4 | 268 | 4661046 | 121 | |
BG717628.1 | - | 4 - 5 | 508 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
