Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006012.2 | NP_006003.1 | 4 - 5 | 680 | 73 | |
| GenBank mRNA | AK311973.1 | - | 4 - 5 | 580 | 73 | |
| BC002956.1 | - | 4 - 5 | 680 | 3542292 | 73 | |
| Z50853.1 | - | 4 - 5 | 576 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000112.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001722.hg.1 | ENST00000245816.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001722.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001722.hg.1 | TC19000112.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000594780.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006795.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | NM_006012.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | TC1900006795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000245816.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596149.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000596605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | ENST00000597326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900147715.hg.1 | NM_006012.2 | Refseq |

CLPP
Genomic Map