Assay Details
| Gene Symbol: HADHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000182.4 | NP_000173.2 | 7 - 8 | 811 | 60 | |
| GenBank mRNA | AK293398.1 | - | 4 - 5 | 304 | 60 | |
| AK302532.1 | - | 6 - 7 | 558 | 60 | ||
| AK313027.1 | - | 7 - 8 | 704 | 60 | ||
| BC009235.2 | - | 7 - 8 | 706 | 2966432 | 60 | |
| D16480.1 | - | 7 - 8 | 708 | 60 | ||
| FJ626228.1 | - | 6 - 7 | 547 | 60 | ||
| GU727642.1 | - | 7 - 8 | 706 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001665.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
response to insulin
cardiolipin acyl-chain remodeling
response to drug
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acetyltransferase activity
enoyl-CoA hydratase activity
protein binding
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding
fatty acid beta-oxidation
response to insulin
cardiolipin acyl-chain remodeling
response to drug
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acetyltransferase activity
enoyl-CoA hydratase activity
protein binding
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026670.hg.1 | TC02001665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026670.hg.1 | ENST00000380649.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026670.hg.1 | ENST00000457468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026670.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026669.hg.1 | TC02001665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026669.hg.1 | ENST00000380649.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026669.hg.1 | ENST00000457468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026669.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026671.hg.1 | TC02001665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026671.hg.1 | ENST00000380649.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026671.hg.1 | ENST00000457468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02026671.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187542.hg.1 | TC0200012030.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187543.hg.1 | TC0200012030.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200187544.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187544.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187544.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200187544.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200187544.hg.1 | TC0200012030.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000380649.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000461025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000471743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | ENST00000492433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012030.hg.1 | NM_000182.4 | Refseq |

HADHA
Genomic Map