Assay Details
| Gene Symbol: PTPRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002850.3 | NP_002841.3 | 26 - 27 | 4334 | 108 | |
| XM_005259600.2 | XP_005259657.1 | 23 - 24 | 4263 | 108 | ||
| XM_011528157.2 | XP_011526459.1 | 17 - 18 | 2879 | 108 | ||
| XM_011528158.2 | XP_011526460.1 | 11 - 12 | 1742 | 108 | ||
| XM_017027065.1 | XP_016882554.1 | 24 - 25 | 4275 | 108 | ||
| XM_017027066.1 | XP_016882555.1 | 24 - 25 | 4293 | 108 | ||
| XM_017027067.1 | XP_016882556.1 | 24 - 25 | 4191 | 108 | ||
| XM_017027068.1 | XP_016882557.1 | 23 - 24 | 4263 | 108 | ||
| XM_017027069.1 | XP_016882558.1 | 23 - 24 | 4248 | 108 | ||
| XM_017027073.1 | XP_016882562.1 | 18 - 19 | 3478 | 108 | ||
| XM_017027074.1 | XP_016882563.1 | 19 - 20 | 3020 | 108 | ||
| XM_017027075.1 | XP_016882564.1 | 18 - 19 | 3008 | 108 | ||
| GenBank mRNA | AB209333.1 | - | 18 - 19 | 3041 | 108 | |
| U35234.1 | - | 25 - 26 | 4208 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PTPRS
Genomic Map