Assay Details
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001484.2 | NP_001001484.1 | 4 - 5 | 984 | 139 | |
NM_001261836.1 | NP_001248765.1 | 3 - 4 | 850 | 139 | ||
NM_001261838.1 | NP_001248767.1 | 2 - 3 | 370 | 139 | ||
NM_030664.4 | NP_109589.2 | 4 - 5 | 929 | 139 | ||
XM_017016927.1 | XP_016872416.1 | 3 - 4 | 768 | 139 | ||
GenBank mRNA | AK314910.1 | - | 3 - 4 | 850 | 139 | |
BC015092.1 | - | 4 - 5 | 907 | 4049884 | 139 | |
BC050411.1 | - | 4 - 5 | 957 | 6472327 | 139 | |
BU172463.1 | - | 2 - 3 | 317 | 139 | ||
BX648556.1 | - | 3 - 4 | 822 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000122.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | ENST00000298942.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | ENST00000343656.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | ENST00000378000.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | ENST00000535784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10001348.hg.1 | TC10000122.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147115.hg.1 | TC1000006880.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000147127.hg.1 | TC1000006880.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
