Assay Details
| Gene Symbol: ARHGAP39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308207.1 | NP_001295136.1 | 8 - 9 | 3146 | 78 | |
| NM_001308208.1 | NP_001295137.1 | 7 - 8 | 2799 | 78 | ||
| NM_025251.2 | NP_079527.1 | 7 - 8 | 2765 | 78 | ||
| XM_011517308.1 | XP_011515610.1 | 8 - 9 | 3046 | 78 | ||
| XM_011517309.1 | XP_011515611.1 | 8 - 9 | 2950 | 78 | ||
| XM_011517312.2 | XP_011515614.1 | 6 - 7 | 3584 | 78 | ||
| XM_017013870.1 | XP_016869359.1 | 9 - 10 | 4118 | 78 | ||
| XM_017013871.1 | XP_016869360.1 | 8 - 9 | 3177 | 78 | ||
| GenBank mRNA | AB051475.1 | - | 8 - 9 | 3150 | 78 | |
| AK092501.1 | - | 3 - 4 | 212 | 78 | ||
| AK296353.1 | - | 7 - 8 | 2796 | 78 | ||
| AK303851.1 | - | 4 - 5 | 1468 | 78 | ||
| BC013071.2 | - | 1 - 2 | 76 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001758.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08023398.hg.1 | TC08001758.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023398.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023398.hg.1 | ENST00000377307.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023398.hg.1 | ENST00000540274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023398.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | TC08001758.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | ENST00000377307.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | ENST00000540274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023397.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183992.hg.1 | TC0800012225.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000276826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_001308208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012225.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183994.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183994.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183994.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183994.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183994.hg.1 | TC0800012225.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | ENST00000377307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | ENST00000528810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | NM_025251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183995.hg.1 | TC0800012225.hg.1 | Clariom D TC |

ARHGAP39
Genomic Map