Assay Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006530.3 | NP_006521.1 | 3 - 4 | 502 | 150 | |
| GenBank mRNA | AJ245746.1 | - | 3 - 4 | 450 | 150 | |
| AK001413.1 | - | 3 - 4 | 275 | 150 | ||
| BC000994.2 | - | 3 - 4 | 363 | 2899965 | 150 | |
| U61384.1 | - | 3 - 4 | 460 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cytoskeleton organization
mitotic nuclear division
regulation of growth
histone H4 acetylation
histone H2A acetylation
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
structural constituent of cytoskeleton
protein binding
protein C-terminus binding
transcription, DNA-templated
cytoskeleton organization
mitotic nuclear division
regulation of growth
histone H4 acetylation
histone H2A acetylation
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
structural constituent of cytoskeleton
protein binding
protein C-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000548020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552949.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_001300950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008134.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000247843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000549685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | ENST00000552955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | NM_006530.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159614.hg.1 | TC1200008134.hg.1 | Clariom D TC |

YEATS4
Genomic Map