Assay Details
| Gene Symbol: IGSF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032880.4 | NP_116269.3 | 7 - 8 | 1486 | 56 | |
| XM_011542319.2 | XP_011540621.1 | 5 - 6 | 852 | 56 | ||
| XM_017002604.1 | XP_016858093.1 | 7 - 8 | 1132 | 56 | ||
| XM_017002605.1 | XP_016858094.1 | 6 - 7 | 949 | 56 | ||
| GenBank mRNA | AK123172.1 | - | 3 - 4 | 1427 | 56 | |
| BC007618.1 | - | 7 - 8 | 1406 | 3355289 | 56 | |
| BF939845.1 | - | 3 - 4 | 56 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000239.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01003822.hg.1 | TC01000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003822.hg.1 | ENST00000251296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003822.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003823.hg.1 | TC01000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003823.hg.1 | ENST00000251296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003823.hg.1 | ENST00000497331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003823.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149764.hg.1 | TC0100007136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149765.hg.1 | TC0100007136.hg.1 | Clariom D TC |

IGSF21
Genomic Map