Assay Details
| Gene Symbol: TGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304430.1 | NP_001291359.1 | 9 - 10 | 1041 | 108 | |
| NM_014305.3 | NP_055120.1 | 9 - 10 | 965 | 108 | ||
| NR_130731.1 | 9 - 10 | 961 | 108 | |||
| NR_130732.1 | 8 - 9 | 874 | 108 | |||
| XM_011521065.2 | XP_011519367.1 | 9 - 10 | 968 | 108 | ||
| XM_011521066.2 | XP_011519368.1 | 9 - 10 | 849 | 108 | ||
| GenBank mRNA | AF048686.1 | - | 9 - 10 | 867 | 108 | |
| AJ006068.1 | - | 9 - 10 | 919 | 108 | ||
| AK313251.1 | - | 9 - 10 | 875 | 108 | ||
| AY544125.1 | - | 9 - 10 | 842 | 108 | ||
| BC005284.1 | - | 9 - 10 | 947 | 108 | ||
| BC033675.1 | - | 9 - 10 | 894 | 5175390 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13008901.hg.1 | ENST00000261296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008901.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008901.hg.1 | TC13000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163504.hg.1 | TC1300009492.hg.1 | Clariom D TC |

TGDS
Genomic Map