Assay Details
| Gene Symbol: PANX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF398509.1 | - | 4 - 5 | 1588 | 97 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000227638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | ENST00000436171.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008797.hg.1 | NM_015368.3 | Refseq |

PANX1
Genomic Map