| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17015247.hg.1 |
ENST00000252172.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17015247.hg.1 |
NM_003802.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17015247.hg.1 |
TC17001130.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000252172.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000418404.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000621918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
NM_003802.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000252172.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000418404.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
ENST00000621918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
NM_003802.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700176014.hg.1 |
TC1700009740.hg.1 |
Clariom D TC |