Assay Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081686.1 | NP_001075155.1 | 1 - 2 | 311 | 97 | |
| NM_021176.2 | NP_066999.1 | 1 - 2 | 311 | 97 | ||
| XM_011511564.2 | XP_011509866.1 | 1 - 2 | 328 | 97 | ||
| GenBank mRNA | BC104778.1 | - | 1 - 2 | 311 | 8143781 | 97 |
| BC113376.1 | - | 1 - 2 | 311 | 8322428 | 97 | |
| BQ268961.1 | - | 1 - 2 | 246 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000999.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014606.hg.1 | TC02000999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014606.hg.1 | ENST00000375363.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014606.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014606.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | TC02000999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | ENST00000375363.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014609.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014608.hg.1 | TC02000999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014608.hg.1 | ENST00000375363.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014608.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014608.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014608.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014610.hg.1 | TC02000999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014610.hg.1 | ENST00000375363.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014610.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014610.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014610.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169144.hg.1 | TC0200009903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | ENST00000461586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009903.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169141.hg.1 | TC0200009903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169139.hg.1 | TC0200009903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000282075.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000375363.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000421979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | ENST00000429379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | NM_001081686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | NM_021176.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169143.hg.1 | TC0200009903.hg.1 | Clariom D TC |

G6PC2
Genomic Map