Assay Details
| Gene Symbol: FN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306129.1 | NP_001293058.1 | 23 - 24 | 3873 | 97 | |
| NM_001306130.1 | NP_001293059.1 | 23 - 24 | 3873 | 97 | ||
| NM_001306131.1 | NP_001293060.1 | 23 - 24 | 3873 | 97 | ||
| NM_001306132.1 | NP_001293061.1 | 23 - 24 | 3873 | 97 | ||
| NM_002026.3 | NP_002017.1 | 23 - 24 | 3871 | 97 | ||
| NM_212474.2 | NP_997639.1 | 23 - 24 | 3871 | 97 | ||
| NM_212476.2 | NP_997641.1 | 23 - 24 | 3871 | 97 | ||
| NM_212478.2 | NP_997643.1 | 23 - 24 | 3871 | 97 | ||
| NM_212482.2 | NP_997647.1 | 23 - 24 | 3871 | 97 | ||
| XM_005246397.1 | XP_005246454.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246398.1 | XP_005246455.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246399.1 | XP_005246456.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246401.1 | XP_005246458.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246402.1 | XP_005246459.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246403.1 | XP_005246460.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246404.1 | XP_005246461.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246405.1 | XP_005246462.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246406.1 | XP_005246463.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246407.1 | XP_005246464.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246408.1 | XP_005246465.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246409.1 | XP_005246466.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246410.1 | XP_005246467.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246411.1 | XP_005246468.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246412.1 | XP_005246469.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246414.1 | XP_005246471.1 | 23 - 24 | 3970 | 97 | ||
| XM_005246416.1 | XP_005246473.1 | 23 - 24 | 3970 | 97 | ||
| XM_017003692.1 | XP_016859181.1 | 23 - 24 | 3970 | 97 | ||
| XM_017003693.1 | XP_016859182.1 | 23 - 24 | 3970 | 97 | ||
| XM_017003694.1 | XP_016859183.1 | 23 - 24 | 3970 | 97 | ||
| XM_017003695.1 | XP_016859184.1 | 23 - 24 | 3970 | 97 | ||
| GenBank mRNA | AB191261.1 | - | 23 - 24 | 3871 | 97 | |
| AB209840.1 | - | 23 - 24 | 3863 | 97 | ||
| AK300458.1 | - | 2 - 3 | 219 | 97 | ||
| AL832202.1 | - | 23 - 24 | 3872 | 97 | ||
| AL832771.1 | - | 23 - 24 | 3872 | 97 | ||
| BC117176.1 | - | 23 - 24 | 3851 | 40125727 | 97 | |
| BC143754.1 | - | 23 - 24 | 3851 | 9052268 | 97 | |
| BC143763.1 | - | 23 - 24 | 3851 | 9052277 | 97 | |
| BX537590.1 | - | 23 - 24 | 3872 | 97 | ||
| BX538017.1 | - | 23 - 24 | 3872 | 97 | ||
| BX538018.1 | - | 23 - 24 | 3872 | 97 | ||
| BX640608.1 | - | 23 - 24 | 3871 | 97 | ||
| BX640731.1 | - | 23 - 24 | 3872 | 97 | ||
| BX640803.1 | - | 23 - 24 | 3871 | 97 | ||
| BX640875.1 | - | 23 - 24 | 3872 | 97 | ||
| BX640920.1 | - | 3 - 4 | 307 | 97 | ||
| BX640999.1 | - | 18 - 19 | 2847 | 97 | ||
| CR749281.1 | - | 23 - 24 | 3910 | 97 | ||
| CR749316.1 | - | 23 - 24 | 3870 | 97 | ||
| CR749317.1 | - | 23 - 24 | 3870 | 97 | ||
| CR749666.1 | - | 3 - 4 | 486 | 97 | ||
| U42457.1 | - | 2 - 3 | 261 | 97 | ||
| X02761.1 | - | 23 - 24 | 3525 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002747.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300458.1,CR749666.1 transcripts.
Set Membership: |
|

FN1
Genomic Map