Assay Details
| Gene Symbol: PXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006719532.1 | XP_006719595.1 | 7 | 1269 | 60 | |
| XM_006719534.1 | XP_006719597.1 | 7 | 1424 | 60 | ||
| XM_006719535.3 | XP_006719598.1 | 8 | 1352 | 60 | ||
| XM_006719536.2 | XP_006719599.1 | 7 | 1241 | 60 | ||
| XM_011538622.1 | XP_011536924.1 | 8 | 1351 | 60 | ||
| XM_011538623.1 | XP_011536925.1 | 8 | 1538 | 60 | ||
| XM_017019726.1 | XP_016875215.1 | 7 | 1837 | 60 | ||
| XM_017019727.1 | XP_016875216.1 | 7 | 1637 | 60 | ||
| XM_017019728.1 | XP_016875217.1 | 7 | 1628 | 60 | ||
| XM_017019729.1 | XP_016875218.1 | 7 | 1837 | 60 | ||
| XM_017019730.1 | XP_016875219.1 | 7 | 1837 | 60 | ||
| XM_017019731.1 | XP_016875220.1 | 8 | 1722 | 60 | ||
| XM_017019732.1 | XP_016875221.1 | 7 | 1837 | 60 | ||
| XM_017019733.1 | XP_016875222.1 | 7 | 1837 | 60 | ||
| XM_017019734.1 | XP_016875223.1 | 7 | 1149 | 60 | ||
| XM_017019735.1 | XP_016875224.1 | 7 | 1837 | 60 | ||
| GenBank mRNA | AB209034.1 | - | 7 | 1247 | 60 | |
| AK026695.1 | - | 1 | 161 | 60 | ||
| BC052611.1 | - | 1 | 353 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PXN
Genomic Map