Assay Details
| Gene Symbol: ECSCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077693.3 | NP_001071161.1 | 1 - 2 | 140 | 62 | |
| NM_001293739.1 | NP_001280668.1 | 1 - 2 | 140 | 62 | ||
| NR_121659.1 | 1 - 2 | 140 | 62 | |||
| GenBank mRNA | BC146912.1 | - | 1 - 2 | 112 | 9056840 | 62 |
| BC146913.1 | - | 1 - 2 | 112 | 9056841 | 62 | |
| BI834795.1 | - | 1 - 2 | 90 | 62 | ||
| DQ462572.1 | - | 1 - 2 | 134 | 62 | ||
| EU025066.1 | - | 1 - 2 | 140 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05003431.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025299.hg.1 | TC05003431.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025299.hg.1 | ENST00000515823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025299.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025300.hg.1 | TC05003431.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025300.hg.1 | ENST00000515823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025300.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196901.hg.1 | TC0500013372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196902.hg.1 | TC0500013372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500196900.hg.1 | TC0500013372.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | ENST00000618155.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001077693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NM_001293739.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013372.hg.1 | NR_121659.1 | Refseq |

ECSCR
Genomic Map