Assay Details
| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174916.2 | NP_777576.1 | 44 - 45 | 4906 | 69 | |
| GenBank mRNA | AF525401.1 | - | 44 - 45 | 4789 | 69 | |
| AK127478.1 | - | 8 - 9 | 761 | 69 | ||
| AY061886.1 | - | 44 - 45 | 4840 | 69 | ||
| BC113505.1 | - | 44 - 45 | 4918 | 8322557 | 69 | |
| BC113507.1 | - | 44 - 45 | 4918 | 8322559 | 69 | |
| BX648400.1 | - | 16 - 17 | 1718 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001271.hg.1
Set Membership: |
|

UBR1
Genomic Map