Assay Details
| Gene Symbol: RGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080496.2 | NP_001073965.2 | 4 - 5 | 486 | 61 | |
| GenBank mRNA | AK289566.1 | - | 4 - 5 | 479 | 61 | |
| BC001725.1 | - | 4 - 5 | 414 | 3533857 | 61 | |
| D87447.1 | - | 4 - 5 | 423 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000193.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09002177.hg.1 | TC09000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002177.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002177.hg.1 | ENST00000456972.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002177.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007098.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | ENST00000378078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | ENST00000496906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | NM_001080496.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148258.hg.1 | TC0900007098.hg.1 | Clariom D TC |

RGP1
Genomic Map