Assay Details
| Gene Symbol: MND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253861.1 | NP_001240790.1 | 2 - 3 | 156 | 82 | |
| NM_032117.3 | NP_115493.1 | 2 - 3 | 156 | 82 | ||
| NR_045605.1 | 2 - 3 | 156 | 82 | |||
| XM_005263275.2 | XP_005263332.1 | 2 - 3 | 115 | 82 | ||
| XM_017008688.1 | XP_016864177.1 | 2 - 3 | 513 | 82 | ||
| GenBank mRNA | AK312172.1 | - | 2 - 3 | 115 | 82 | |
| AY028916.1 | - | 2 - 3 | 95 | 82 | ||
| BC032142.2 | - | 2 - 3 | 110 | 5095005 | 82 | |
| BF975093.1 | - | 2 - 3 | 125 | 82 | ||
| BG613292.1 | - | 2 - 3 | 118 | 82 | ||
| BG758047.1 | - | 2 - 3 | 125 | 82 | ||
| BM833450.1 | - | 2 - 3 | 156 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000773.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017008688.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | TC04000773.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | ENST00000240488.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | ENST00000504860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010618.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000578990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000418249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009050.hg.1 | ENST00000602631.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000578990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009037.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000240488.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000503967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000504860.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000508731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000509752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | ENST00000622785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NM_001253861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NM_032117.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | NR_045605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161997.hg.1 | TC0400009037.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 192975 | - | - |

MND1
Genomic Map