Assay Details
| Gene Symbol: PRRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286808.1 | NP_001273737.1 | 7 - 8 | 1214 | 76 | |
| NM_130809.4 | NP_570721.1 | 7 - 8 | 1214 | 76 | ||
| XM_006714529.3 | XP_006714592.1 | 7 - 8 | 1422 | 76 | ||
| GenBank mRNA | AJ427339.1 | - | 7 - 8 | 1167 | 76 | |
| AJ441110.3 | - | 7 - 8 | 1167 | 76 | ||
| AJ515158.1 | - | 7 - 8 | 1167 | 76 | ||
| AJ515428.1 | - | 7 - 8 | 1167 | 76 | ||
| AJ515429.1 | - | 7 - 8 | 1167 | 76 | ||
| AK057437.1 | - | 7 - 8 | 1151 | 76 | ||
| AK090486.1 | - | 6 - 7 | 1039 | 76 | ||
| AL831963.1 | - | 7 - 8 | 1176 | 76 | ||
| AL832056.1 | - | 7 - 8 | 1177 | 76 | ||
| AL832600.2 | - | 7 - 8 | 1173 | 76 | ||
| BC017066.1 | - | 7 - 8 | 1151 | 3913311 | 76 | |
| BX099255.1 | - | 4 - 5 | 410 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000608.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | TC05000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000296666.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000330542.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000442138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000512535.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05008458.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500158728.hg.1 | TC0500008558.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000296666.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000442138.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000507774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512635.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000512871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | ENST00000513427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_001286808.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008558.hg.1 | NM_130809.4 | Refseq |

PRRC1
Genomic Map