Assay Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164114.1 | NP_001157586.1 | 3 - 4 | 599 | 97 | |
| NM_001164115.1 | NP_001157587.1 | 3 - 4 | 761 | 97 | ||
| NM_001164116.1 | NP_001157588.1 | 4 - 5 | 785 | 97 | ||
| NM_020356.3 | NP_065089.2 | 3 - 4 | 761 | 97 | ||
| XM_006723831.3 | XP_006723894.1 | 4 - 5 | 1185 | 97 | ||
| GenBank mRNA | AJ276678.1 | - | 4 - 5 | 785 | 97 | |
| AK027760.1 | - | 3 - 4 | 758 | 97 | ||
| AK295613.1 | - | 3 - 4 | 464 | 97 | ||
| AK308057.1 | - | 3 - 4 | 762 | 97 | ||
| AK314653.1 | - | 3 - 4 | 694 | 97 | ||
| BC027951.1 | - | 3 - 4 | 711 | 5205865 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000429.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | ENST00000360314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | ENST00000371336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | ENST00000434344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005953.hg.1 | TC20000429.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | TC2000007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154282.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |

CASS4
Genomic Map