Assay Details
| Gene Symbol: OLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172632.1 | NP_001166103.1 | 2 - 3 | 294 | 77 | |
| NM_001172633.1 | NP_001166104.1 | 2 - 3 | 294 | 77 | ||
| NM_002543.3 | NP_002534.1 | 2 - 3 | 294 | 77 | ||
| GenBank mRNA | AB010710.1 | - | 2 - 3 | 241 | 77 | |
| AB102861.1 | - | 2 - 3 | 180 | 77 | ||
| AF035776.1 | - | 2 - 3 | 226 | 77 | ||
| AK292124.1 | - | 2 - 3 | 235 | 77 | ||
| AK295409.1 | - | 2 - 3 | 235 | 77 | ||
| AL551100.3 | - | 2 - 3 | 218 | 77 | ||
| AX429248.1 | - | 2 - 3 | 241 | 77 | ||
| BC022295.1 | - | 2 - 3 | 235 | 4722086 | 77 | |
| BP219342.1 | - | 2 - 3 | 294 | 77 | ||
More Information
Set Membership: |
|

OLR1
Genomic Map