Assay Details
| Gene Symbol: LRRC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001887.1 | XP_016857376.1 | 21 - 22 | 4225 | 109 | |
| XM_017001890.1 | XP_016857379.1 | 21 - 22 | 5472 | 109 | ||
| XM_017001891.1 | XP_016857380.1 | 22 - 23 | 5549 | 109 | ||
| XM_017001892.1 | XP_016857381.1 | 21 - 22 | 4225 | 109 | ||
| XM_017001893.1 | XP_016857382.1 | 21 - 22 | 4021 | 109 | ||
| XM_017001894.1 | XP_016857383.1 | 22 - 23 | 4067 | 109 | ||
| GenBank mRNA | AB037786.1 | - | 3 - 4 | 1931 | 109 | |
More Information
Set Membership: |
|

LRRC7
Genomic Map