Assay Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164116.1 | NP_001157588.1 | 1 - 2 | 99 | 129 | |
| XM_006723831.3 | XP_006723894.1 | 1 - 2 | 661 | 129 | ||
| GenBank mRNA | AJ276678.1 | - | 1 - 2 | 99 | 129 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000429.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ276678.1,NM_001164116.1,XM_006723831.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005945.hg.1 | ENST00000360314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005945.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005945.hg.1 | TC20000429.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | ENST00000360314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | ENST00000371336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | ENST00000434344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005949.hg.1 | TC20000429.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | TC2000007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154277.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000434344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | ENST00000497244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164115.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007820.hg.1 | NM_020356.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154273.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154273.hg.1 | NM_001164116.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000154273.hg.1 | TC2000007820.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000154273.hg.1 | ENST00000360314.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000154273.hg.1 | NM_001164116.1 | Refseq |

CASS4
Genomic Map