Assay Details
| Gene Symbol: SCAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021228.2 | NP_067051.2 | 1 - 2 | 118 | 112 | |
| GenBank mRNA | AK024444.1 | - | 1 - 2 | 118 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000742.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK024444.1,NM_021228.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19010375.hg.1 | ENST00000360565.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010375.hg.1 | ENST00000447618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010375.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010375.hg.1 | TC19000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010373.hg.1 | ENST00000360565.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010373.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010373.hg.1 | TC19000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008549.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166251.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166251.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166251.hg.1 | TC1900008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166251.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166251.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | NM_021228.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | TC1900008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000360565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000595242.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000598359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | ENST00000601038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166257.hg.1 | NM_021228.2 | Refseq |

SCAF1
Genomic Map