Assay Details
| Gene Symbol: FUZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527345.1 | XP_011525647.1 | 2 - 3 | 370 | 96 | |
| XM_011527347.1 | XP_011525649.1 | 3 - 4 | 328 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527345.1, XM_011527347.1 transcripts.
Set Membership: |
|

FUZ
Genomic Map