Assay Details
| Gene Symbol: IQGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178229.4 | NP_839943.2 | 36 - 37 | 4744 | 72 | |
| XM_011509198.2 | XP_011507500.1 | 36 - 37 | 4733 | 72 | ||
| XM_011509200.2 | XP_011507502.1 | 25 - 26 | 3913 | 72 | ||
| XM_011509201.2 | XP_011507503.1 | 24 - 25 | 3302 | 72 | ||
| XM_017000317.1 | XP_016855806.1 | 36 - 37 | 4730 | 72 | ||
| XM_017000318.1 | XP_016855807.1 | 34 - 35 | 4487 | 72 | ||
| GenBank mRNA | AB105103.1 | - | 36 - 37 | 4680 | 72 | |
| AK122653.1 | - | 22 - 23 | 2952 | 72 | ||
| AY253300.1 | - | 36 - 37 | 4669 | 72 | ||
| BC131536.1 | - | 36 - 37 | 4702 | 40082932 | 72 | |
| BM910515.1 | - | 2 - 3 | 221 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003343.hg.1
Set Membership: |
|

IQGAP3
Genomic Map