Assay Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135254.1 | NP_001128726.1 | 2 - 3 | 920 | 107 | |
| NM_002584.2 | NP_002575.1 | 2 - 3 | 920 | 107 | ||
| NM_013945.2 | NP_039236.1 | 2 - 3 | 920 | 107 | ||
| GenBank mRNA | BC121165.2 | - | 2 - 3 | 402 | 40121582 | 107 |
| BC121166.2 | - | 2 - 3 | 402 | 40121584 | 107 | |
| DQ322591.1 | - | 2 - 3 | 405 | 107 | ||
| X96743.1 | - | 2 - 3 | 921 | 107 | ||
| Z35141.1 | - | 2 - 3 | 424 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000243.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
anatomical structure morphogenesis
regulation of cell fate commitment
skeletal muscle satellite cell commitment
spinal cord association neuron differentiation
dorsal/ventral neural tube patterning
positive regulation of histone methylation
negative regulation of apoptotic process
regulation of protein binding
skeletal muscle tissue regeneration
positive regulation of transcription from RNA polymerase II promoter
neuron fate commitment
embryonic skeletal system development
cartilage development
muscle tissue morphogenesis
positive regulation of myoblast proliferation
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
chromatin remodeling
transcription, DNA-templated
anatomical structure morphogenesis
regulation of cell fate commitment
skeletal muscle satellite cell commitment
spinal cord association neuron differentiation
dorsal/ventral neural tube patterning
positive regulation of histone methylation
negative regulation of apoptotic process
regulation of protein binding
skeletal muscle tissue regeneration
positive regulation of transcription from RNA polymerase II promoter
neuron fate commitment
embryonic skeletal system development
cartilage development
muscle tissue morphogenesis
positive regulation of myoblast proliferation
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | TC01000243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | ENST00000375375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | ENST00000420770.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003844.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149801.hg.1 | TC0100007147.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000375375.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000400661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | ENST00000420770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_001135254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_002584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007147.hg.1 | NM_013945.2 | Refseq |

PAX7
Genomic Map