| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
ENST00000261641.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
ENST00000315321.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002774.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002775.hg.1 |
ENST00000261641.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002775.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002775.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002775.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002775.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
ENST00000261641.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
ENST00000315321.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002776.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002777.hg.1 |
ENST00000315321.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002777.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002777.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002777.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002777.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002778.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002778.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002778.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002778.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002781.hg.1 |
ENST00000315321.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002781.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002781.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002781.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002781.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002779.hg.1 |
ENST00000315321.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002779.hg.1 |
ENST00000353383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002779.hg.1 |
ENST00000395776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002779.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002779.hg.1 |
TC17000201.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700149883.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149883.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149883.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149883.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149883.hg.1 |
TC1700007080.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149884.hg.1 |
TC1700007080.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149885.hg.1 |
TC1700007080.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000353383.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000395774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000471135.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000489697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000582514.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
ENST00000583166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007080.hg.1 |
NM_030665.3 |
Refseq |