Assay Details
| Gene Symbol: GABBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005458.7 | NP_005449.5 | 1 - 2 | 781 | 120 | |
| GenBank mRNA | AF056085.1 | - | 1 - 2 | 778 | 120 | |
| AF069755.1 | - | 1 - 2 | 477 | 120 | ||
| AF074483.1 | - | 1 - 2 | 321 | 120 | ||
| AF095784.1 | - | 1 - 2 | 452 | 120 | ||
| AF099033.1 | - | 1 - 2 | 321 | 120 | ||
| AJ012188.1 | - | 1 - 2 | 321 | 120 | ||
| BC035071.2 | - | 2 - 3 | 616 | 5260138 | 120 | |
| JQ308173.1 | - | 1 - 2 | 321 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001406.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09017548.hg.1 | TC09001406.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017548.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017548.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017551.hg.1 | TC09001406.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017551.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017551.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017549.hg.1 | TC09001406.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017549.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017549.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017550.hg.1 | TC09001406.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017550.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017550.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174819.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174819.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174819.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174819.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174819.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174817.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174817.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174817.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174817.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174817.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | ENST00000477471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010975.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174818.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174818.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174818.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174818.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174818.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174821.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174821.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174821.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174821.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174821.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174820.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174820.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174820.hg.1 | ENST00000259455.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174820.hg.1 | NM_005458.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174820.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174822.hg.1 | TC0900010975.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148792 | - | - |

GABBR2
Genomic Map