Assay Details
| Gene Symbol: KCNQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172107.3 | NP_742105.1 | 11 - 12 | 1465 | 117 | |
| GenBank mRNA | AF033348.1 | - | 11 - 12 | 1378 | 117 | |
More Information
Set Membership: |
|

KCNQ2
Genomic Map