Assay Details
| Gene Symbol: RMDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170791.2 | NP_001164262.1 | 2 - 3 | 567 | 107 | |
| NM_001170792.2 | NP_001164263.1 | 2 - 3 | 638 | 107 | ||
| NM_001322211.1 | NP_001309140.1 | 2 - 3 | 636 | 107 | ||
| NM_001322212.1 | NP_001309141.1 | 2 - 3 | 567 | 107 | ||
| XM_011532616.2 | XP_011530918.1 | 2 - 3 | 539 | 107 | ||
| XM_011532617.2 | XP_011530919.1 | 2 - 3 | 625 | 107 | ||
| XM_011532618.2 | XP_011530920.1 | 3 - 4 | 749 | 107 | ||
| XM_011532619.2 | XP_011530921.1 | 2 - 3 | 632 | 107 | ||
| XM_017003476.1 | XP_016858965.1 | 2 - 3 | 539 | 107 | ||
| XM_017003477.1 | XP_016858966.1 | 2 - 3 | 631 | 107 | ||
| XM_017003478.1 | XP_016858967.1 | 2 - 3 | 539 | 107 | ||
| GenBank mRNA | AY358269.1 | - | 2 - 3 | 567 | 107 | |
| BC024243.2 | - | 2 - 3 | 640 | 4798767 | 107 | |
More Information
Set Membership: |
|

RMDN2
Genomic Map