Assay Details
| Gene Symbol: COL5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015719.3 | NP_056534.2 | 45 - 46 | 3427 | 117 | |
| XM_011528042.2 | XP_011526344.1 | 45 - 46 | 3485 | 117 | ||
| XM_017026849.1 | XP_016882338.1 | 18 - 19 | 1128 | 117 | ||
| GenBank mRNA | AB209175.1 | - | 43 - 44 | 3080 | 117 | |
| AF177941.1 | - | 45 - 46 | 3427 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001156.hg.1
Set Membership: |
|

COL5A3
Genomic Map