Assay Details
| Gene Symbol: CNGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019098.4 | NP_061971.3 | 16 - 17 | 1976 | 93 | |
| XM_011517138.2 | XP_011515440.1 | 14 - 15 | 1631 | 93 | ||
| GenBank mRNA | AF228520.1 | - | 13 - 14 | 1574 | 93 | |
| AF272900.1 | - | 16 - 17 | 1974 | 93 | ||
| BC150601.1 | - | 16 - 17 | 1954 | 9056971 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001392.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
cation transport
signal transduction
visual perception
regulation of membrane potential
potassium ion transmembrane transport
intracellular cAMP activated cation channel activity
intracellular cGMP activated cation channel activity
voltage-gated potassium channel activity
cGMP binding
transport
cation transport
signal transduction
visual perception
regulation of membrane potential
potassium ion transmembrane transport
intracellular cAMP activated cation channel activity
intracellular cGMP activated cation channel activity
voltage-gated potassium channel activity
cGMP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018026.hg.1 | TC08001392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018026.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08018026.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800174943.hg.1 | TC0800011023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000320005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000517327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | ENST00000519777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011023.hg.1 | NM_019098.4 | Refseq |

CNGB3
Genomic Map